Learn More
Medchemexpress LLC HY-106408A 5mg Medchemexpress, Salfaprodil CAS:916214-57-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY106408A5MG
Encompass_Preferred
Medchemexpress, HY-106408A 5mg Salfaprodil CAS:916214-57-8 Salfaprodil (Neu2000 potassium) is an NR2B-selective and uncompetitive antagonist of N-methyl-D-aspartate (NMDA). Salfaprodil is also a free radical scavenger. Salfaprodil has excellent neuroprotection against NMDA- and free radical-induced cell death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0691
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.