Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salicylate Reagent, Reagents
Salicylate Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CS007400250C
Quantity | 250 mL |
---|
Chemical Identifiers
7732-18-5, 7782-61-8, 7487-94-7 | |
18.02 | |
water |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
O |
Specifications
Orange to Yellow/Orange | |
100°C | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent | |
1.0507 | |
Non-sterile | |
Salicylate Reagent |
7732-18-5, 7782-61-8, 7487-94-7 | |
H2O | |
O | |
18.02 | |
Glass Bottle | |
250 mL | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction