Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylhydroxamic Acid 98.0+%, TCI America™
SDP

Catalog No. S000825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000825G 25 g
S00085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000825G Supplier TCI America Supplier No. S000825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89-73-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00002110
InChI Key HBROZNQEVUILML-UHFFFAOYSA-N
Synonym salicylhydroxamic acid, 2-hydroxybenzohydroxamic acid, salicylohydroxamic acid, sham, salicylohydroximic acid, benzamide, n,2-dihydroxy, 2-hydroxybenzhydroxamic acid, o-hydroxybenzohydroxamic acid, sha, 2,n-dihydroxy-benzamide
PubChem CID 66644
ChEBI CHEBI:45615
IUPAC Name N,2-dihydroxybenzamide
SMILES C1=CC=C(C(=C1)C(=O)NO)O

Specifications

Melting Point 173°C
Quantity 25 g
Formula Weight 153.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Salicylhydroxamic Acid
RTECSNumber VO6870000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.