Learn More
Salicylic Acid, ACS Reagent (2-Hydroxybenzoic Acid), Reagents
Specifications
Specifications
| Melting Point | 159°C |
| Boiling Point | 211°C |
| Color | Colorless/White |
| Quantity | 500 g |
| Packaging | Poly Bottle |
| Physical Form | Solid |
| Specific Gravity | 1.443 |
| Grade | ACS |
| Chemical Name or Material | Salicylic Acid, ACS Reagent (2-Hydroxybenzoic Acid) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.