CAS RN 69-72-7
Salicylic acid, ACS, 99+%
CAS: 69-72-7 Molecular Formula: C7H6O3 Molecular Weight (g/mol): 138.12 MDL Number: MFCD00002439 InChI Key: YGSDEFSMJLZEOE-UHFFFAOYSA-N Synonym: salicylic acid,o-hydroxybenzoic acid,2-carboxyphenol,o-carboxyphenol,rutranex,salonil,retarder w,keralyt,duoplant,freezone PubChem CID: 338 ChEBI: CHEBI:16914 IUPAC Name: 2-hydroxybenzoic acid SMILES: OC(=O)C1=CC=CC=C1O
Salicylic acid, 99%
CAS: 69-72-7 Molecular Formula: C7H6O3 Molecular Weight (g/mol): 138.12 MDL Number: MFCD00002439 InChI Key: YGSDEFSMJLZEOE-UHFFFAOYSA-N Synonym: salicylic acid,o-hydroxybenzoic acid,2-carboxyphenol,o-carboxyphenol,rutranex,salonil,retarder w,keralyt,duoplant,freezone PubChem CID: 338 ChEBI: CHEBI:16914 IUPAC Name: 2-hydroxybenzoic acid SMILES: OC(=O)C1=CC=CC=C1O
Salicylic Acid (Crystalline Powder/Certified ACS), Fisher Chemical
CAS: 69-72-7 Molecular Formula: C7H6O3 Molecular Weight (g/mol): 138.12 MDL Number: MFCD00002439 InChI Key: YGSDEFSMJLZEOE-UHFFFAOYSA-N Synonym: salicylic acid,o-hydroxybenzoic acid,2-carboxyphenol,o-carboxyphenol,rutranex,salonil,retarder w,keralyt,duoplant,freezone PubChem CID: 338 ChEBI: CHEBI:16914 IUPAC Name: 2-hydroxybenzoic acid SMILES: OC(=O)C1=CC=CC=C1O
Salicylic Acid, 99+%
CAS: 69-72-7 Molecular Formula: C7H6O3 Molecular Weight (g/mol): 138.12 MDL Number: MFCD00002439 InChI Key: YGSDEFSMJLZEOE-UHFFFAOYSA-N Synonym: salicylic acid,o-hydroxybenzoic acid,2-carboxyphenol,o-carboxyphenol,rutranex,salonil,retarder w,keralyt,duoplant,freezone PubChem CID: 338 ChEBI: CHEBI:16914 IUPAC Name: 2-hydroxybenzoic acid SMILES: OC(=O)C1=CC=CC=C1O
Salicylic acid, 99+%, ACS reagent
CAS: 69-72-7 Molecular Formula: C7H6O3 Molecular Weight (g/mol): 138.12 MDL Number: MFCD00002439 InChI Key: YGSDEFSMJLZEOE-UHFFFAOYSA-N Synonym: salicylic acid,o-hydroxybenzoic acid,2-carboxyphenol,o-carboxyphenol,rutranex,salonil,retarder w,keralyt,duoplant,freezone PubChem CID: 338 ChEBI: CHEBI:16914 IUPAC Name: 2-hydroxybenzoic acid SMILES: OC(=O)C1=CC=CC=C1O
Salicylic Acid, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
CAS: 69-72-7 Molecular Formula: C7H6O3 Molecular Weight (g/mol): 138.12 MDL Number: MFCD00002439 InChI Key: YGSDEFSMJLZEOE-UHFFFAOYSA-N IUPAC Name: 2-hydroxybenzoic acid SMILES: OC(=O)C1=CC=CC=C1O
Salicylic acid, For ACS analysis, 99.68%, MP Biomedicals™
CAS: 69-72-7 Molecular Formula: C7H6O3 Molecular Weight (g/mol): 138.12 MDL Number: MFCD00002439 InChI Key: YGSDEFSMJLZEOE-UHFFFAOYSA-N Synonym: salicylic acid,o-hydroxybenzoic acid,2-carboxyphenol,o-carboxyphenol,rutranex,salonil,retarder w,keralyt,duoplant,freezone PubChem CID: 338 ChEBI: CHEBI:16914 IUPAC Name: 2-hydroxybenzoic acid SMILES: OC(=O)C1=CC=CC=C1O
Salicylic Acid, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
CAS: 69-72-7 Molecular Formula: C7H6O3 Molecular Weight (g/mol): 138.12 MDL Number: MFCD00002439 InChI Key: YGSDEFSMJLZEOE-UHFFFAOYSA-N IUPAC Name: 2-hydroxybenzoic acid SMILES: OC(=O)C1=CC=CC=C1O
Salicylic Acid, Crystal, USP, 98-102%, Spectrum™ Chemical
CAS: 69-72-7 Molecular Formula: C7H6O3 Molecular Weight (g/mol): 138.12 MDL Number: MFCD00002439 InChI Key: YGSDEFSMJLZEOE-UHFFFAOYSA-N IUPAC Name: 2-hydroxybenzoic acid SMILES: OC(=O)C1=CC=CC=C1O
Salicylic Acid, Powder, USP, 98-102%, Spectrum™ Chemical
CAS: 69-72-7 Molecular Formula: C7H6O3 Molecular Weight (g/mol): 138.12 MDL Number: MFCD00002439 InChI Key: YGSDEFSMJLZEOE-UHFFFAOYSA-N IUPAC Name: 2-hydroxybenzoic acid SMILES: OC(=O)C1=CC=CC=C1O
2-Hydroxybenzoic Acid 99.5+%, TCI America™
CAS: 69-72-7 Molecular Formula: C7H6O3 Molecular Weight (g/mol): 138.12 MDL Number: MFCD00002439 InChI Key: YGSDEFSMJLZEOE-UHFFFAOYSA-N Synonym: salicylic acid,o-hydroxybenzoic acid,2-carboxyphenol,o-carboxyphenol,rutranex,salonil,retarder w,keralyt,duoplant,freezone PubChem CID: 338 ChEBI: CHEBI:16914 IUPAC Name: 2-hydroxybenzoic acid SMILES: OC(=O)C1=CC=CC=C1O
2-Hydroxybenzoic Acid 99.5+%, TCI America™
CAS: 69-72-7 Molecular Formula: C7H6O3 Molecular Weight (g/mol): 138.12 MDL Number: MFCD00002439 InChI Key: YGSDEFSMJLZEOE-UHFFFAOYSA-N Synonym: salicylic acid,o-hydroxybenzoic acid,2-carboxyphenol,o-carboxyphenol,rutranex,salonil,retarder w,keralyt,duoplant,freezone PubChem CID: 338 ChEBI: CHEBI:16914 IUPAC Name: 2-hydroxybenzoic acid SMILES: OC(=O)C1=CC=CC=C1O
Salicylic Acid, ACS Reagent (2-Hydroxybenzoic Acid), Reagents
CAS: 69-72-7 Molecular Formula: C7H6O3 Molecular Weight (g/mol): 138.12 InChI Key: YGSDEFSMJLZEOE-UHFFFAOYSA-N Synonym: 2-Hydroxybenzoic Acid, 2-Carboxyphenol, o-Hydroxybenzoic Acid IUPAC Name: 2-hydroxybenzoic acid SMILES: OC(=O)C1=CC=CC=C1O
HYDRANAL™-Salicylic acid, Solstice
Buffer substance for Karl Fischer titration
- 1
- 2