Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic Acid, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18606144 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-144 6 x 125 g Amber Glass Bottle
18-606-143 125 g Amber Glass Bottle
18-606-147 2.5 kg Poly Pail
18-606-146 6 x 500 g Amber Glass Bottle
18-606-148 4 x 2.5 kg Poly Pail
18-606-149 12 kg Poly Pail
18-606-150 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-606-144 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1005125GMCS6
Only null left
Add to Cart
Add to Cart

S1005, 69-72-7, C7H6O3

Spectrum™ Chemical Salicylic Acid, Crystal, Reagent, ACS is an alpha-hydroxybenzoic acid and a colorless crystalline acid that is organic and commonly used in organic synthesis. It can be found occurring naturally in unripe fruits and vegetables and most commonly in the white willow tree. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 69-72-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002439
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
IUPAC Name 2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC=C1O

Specifications

Melting Point 158°C to 161°C
CAS Max % 1
Percent Purity 99%
Quantity 6 x 125 g
Packaging Amber Glass Bottle
Residue after Ignition 0.0001
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.