Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic Acid, Powder, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18606159 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-159 50 kg Fiber Drum
18-606-155 500 g Amber Glass Bottle
18-606-156 2.5 kg Poly Pail
18-606-157 12 kg Fiber Drum
18-606-158 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-606-159 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SA13050KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SA130, 69-72-7, C7H6O3

Spectrum™ Chemical Salicylic Acid, Powder, USP is a topical application used to treat skin conditions that involve scaling or overgrowth of skin cells such as psoriasis, dandruff, corns, calluses, and warts. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 69-72-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002439
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
IUPAC Name 2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC=C1O

Specifications

CAS Max % 1
Quantity 50 kg
Ignition Residue 0.0005
Loss on Drying 0.005
Packaging Fiber Drum
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.