Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Manganese(II) chloride, 99+%, anhydrous, -80 mesh
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1190°C |
| Melting Point | 650°C |
| Color | Pink |
| Physical Form | Powder |
| Quantity | 50 g |
| Assay Percent Range | 99% min. (Complexometry) |
| Linear Formula | MnCl2 |
| Fieser | 07,222 |
| Merck Index | 15, 5793 |
| Solubility Information | (10% in dilute HCl) Clear solution |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. May cause damage to organs through prolonged or repeated exposure if inhaled. Causes serious eye damage. |
| Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Do not breathe dust/fume/gas/mist/vapors/spray. Do not eat,drink or smoke when using th |
| EINECSNumber | 231-869-6 |
| RTECSNumber | OO9625000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction