Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saligenin, MP Biomedicals™

Catalog No. ICN15657950 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15657950 50 g
ICN15657980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15657950 Supplier MP Biomedicals, Inc Supplier No. 0215657950

Chemical Identifiers

CAS 90-01-7
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00004617
InChI Key CQRYARSYNCAZFO-UHFFFAOYSA-N
Synonym salicyl alcohol, 2-hydroxybenzyl alcohol, saligenin, 2-hydroxymethyl phenol, 2-methylolphenol, o-hydroxybenzyl alcohol, saligenol, benzenemethanol, 2-hydroxy, diathesin, o-methylolphenol
PubChem CID 5146
ChEBI CHEBI:16464
IUPAC Name 2-(hydroxymethyl)phenol
SMILES OCC1=CC=CC=C1O

Specifications

Color White
Quantity 50 g
Formula Weight 124.14
Physical Form Powder
Chemical Name or Material Saligenin
EINECSNumber 201-960-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.