Learn More
Cayman Chemical PtdIns- 3 4 5-P31 2-dIpalm 1mg
SDP
Supplier:  Cayman Chemical 649201 MG
Encompass_Preferred
A synthetic analog of natural PtdIns featuring C16:0 fatty acids at the sn-1 and sn-2 positions; contains the same inositol and DAG stereochemistry as the natural compound
Catalog No. 50-289-5261