Learn More
Med Vet International Sterile Saline Solution 0.9%, 250mL, Each
Supplier:  Med Vet International RXSALINE0.9250V
Encompass_Preferred
For use in replacement therapy of sodium, chloride and water which may become depleted in many diseases in cattle, swine, sheep, goats, horses, dogs and cats. Because this solution is isotonic with body fluids, it may also be used as a solvent or diluent for antibiotics and other pharmaceuticals and biologicals where compatible, and for washing mucous membranes and other tissue surfaces. Each 100 mL contains 0.9 gram of sodium chloride.
Catalog No. 50-282-8802
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.