Learn More
Vernier™ Salinity Standard
SDPSCI_ED
Catalog No. S16247ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Salinity Sensor
Catalog No. For Use With (Equipment)
S16247ND Salinity Sensor
1 options
Catalog No. S16247ND Supplier Vernier™ Supplier No. SALST
Add to Cart
Add to Cart
Includes
Salinity Standard
This 35 ppt standard is used to calibrate the Salinity Sensor. (500 mL)
  • 35 ppt standard
  • Used to calibrate the Salinity Sensor
  • 500 mL bottle
Warranty and Services
5-year limited warranty
Specifications
Description 500 mL
For Use With (Application) Educational use only
For Use With (Equipment) Salinity Sensor