Learn More
Honeywell Salisbury™ 20 cal/cm2 Flash Kits without Gloves
Specifications
Description Includes: Coat, bib overalls, Pro-Hood™, hard hat, safety Glasses and storage bag
Arc Flash Rating 20 cal/cm²
Certifications/Compliance ASTM F1506, NFPA 70E™
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-138-165E
View Documents
Honeywell Salisbury™
SK202XL
2X-Large N/A N/A N/A
19-138-165C
View Documents
Honeywell Salisbury™
SK20L
Large N/A N/A N/A
19-138-165F
View Documents
Honeywell Salisbury™
SK203XL
3X-Large N/A N/A N/A
19-138-165B
View Documents
Honeywell Salisbury™
SK20M
Medium N/A N/A N/A
19-138-165A
View Documents
Honeywell Salisbury™
SK20S
Small N/A N/A N/A
Description
Description
Specifications
Specifications
Includes: Coat, bib overalls, Pro-Hood™, hard hat, safety Glasses and storage bag
20 cal/cm²
ASTM F1506, NFPA 70E™
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.