Specifications
| Arc Flash Rating | 31 cal/cm² |
| Description | Includes: Coat, bib overalls, Pro-Hood™, hard hat, safety Glasses and storage bag |
| Size | Large |
| Certifications/Compliance | ASTM F1506, NFPA 70E™ |