Learn More
Honeywell Salisbury™ 75 cal/cm2 Flash Kits without Gloves
Supplier: Honeywell Salisbury™ SK75L
This item is not returnable.
View return policy
Description
Specifications
75 cal/cm² | |
ASTM F1506, NFPA 70E™ | |
Large | |
Includes: Coat, bib overalls, Pro-Hood™, hard hat, safety Glasses and storage bag |