Learn More
Honeywell Salisbury™ 75 cal/cm2 Flash Kits without Gloves
Supplier: Honeywell Salisbury™ SK75XL
This item is not returnable.
View return policy
Description
Specifications
75 cal/cm² | |
Includes: Coat, bib overalls, Pro-Hood™, hard hat, safety Glasses and storage bag | |
X-Large | |
ASTM F1506, NFPA 70E™ |