Specifications
| Arc Flash Rating | 12 cal/cm² |
| Description | 42 to 44 in. Chest navy blue coveralls, face shield, hard hat, safety Glasses and Salisbury back pack |
| Material | Cotton |
| Size | Large |
| Certifications/Compliance | NFPA 70E™ 2009, CE 0120 |
| Color | Navy |