Learn More
Honeywell Salisbury™ Coverall Kit
Catalog No. 19139202 Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Arc Flash Rating:
12 cal/cm²
8 cal/cm²
Size:
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Arc Flash Rating Size
19-139-202 8 cal/cm² X-Large
19-139-201 8 cal/cm² Large
19-139-204 12 cal/cm² Large
19-139-206 12 cal/cm² 2X-Large
19-139-205 12 cal/cm² X-Large
19-139-203 8 cal/cm² 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-139-202 Supplier Honeywell Salisbury™ Supplier No. SKCA8XL1200
This item is not returnable. View return policy
Specifications
Arc Flash Rating 8 cal/cm²
Description Arc flash coveralls, AS1200 Hat, hard hat, safety Glasses and Salisbury storage bag
Size X-Large
Certifications/Compliance NFPA 70E™ 2009, CE 0120