Learn More
Honeywell Salisbury™ G99 Portable Glove Inflator Kit - Replacement Parts
Shop All WH Salisbury & Co. Products
Description
- The G99 is a simple, easy to use, portable glove inflator
- Provides a quality means of inspecting gloves in the field
- Glove is secured to the inflator by a nylon strap, and fastened with a hook and pile closure
- Inflation is accomplished by pumping the bellows of the inflator against any surface
- Type I gloves should be inflated no more than 1.5x their normal size
- Type II Salcor™ gloves should be inflated no more than 1.25x their normal size
- Replacement parts help ensure that the pump is working properly whenever and wherever it is needed
Specifications
Specifications
| Color | Red |
| Product Type | Storage Bag |
| For Use With (Equipment) | G99 Portable Glove Inflator Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.