Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Salisbury™ LP Series Low Voltage Cowhide Leather Lineman's Glove Protectors

Catalog No. 18150208 Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Size:
8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-150-208 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-150-208 Supplier Honeywell Salisbury™ Supplier No. ILP10/8
This item is not returnable. View return policy

Glove protectors are 10 in. long and feature a straight cuff with shirred elastic back. Wear over insulating rubber gloves for mechanical protection against cuts, abrasions, and punctures.

  • Constructed of high quality, top grain cowhide leather
  • Straight cuff with shirred elastic back
  • Wear over insulating rubber gloves for mechanical protection
  • Ideal for industrial electrical workers in low voltage situations
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Meets ASTM F696 Specifications
Applications Industrial
Coating Material Uncoated
Color Yellow
Material Top Grain Cowhide Leather
Product Type Glove Protectors
Size 8
For Use With (Application) Mechanical Protection
For Use With (Equipment) Rubber Insulating Gloves
Glove Cuff Style Straight
Series ILP10
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.