Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Salisbury™ Grain Cowhide Leather Linesmen's Work Gloves

Catalog No. 19025774 Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-025774 Small
19-025775 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-025774 Supplier Honeywell Salisbury™ Supplier No. LW2SPE/SM
This item is not returnable. View return policy

Designed specifically for use by lineman, these gloves are constructed of high quality grain cowhide leather for protection against cuts and abrasions.

  • Designed specifically for use by linemen
  • Gloves offer many of the quality features found in leather protectors
  • Grain cowhide leather is soft and flexible, while still being highly resistant to cuts and abrasions
  • Natural pigskin leather cuff
TRUSTED_SUSTAINABILITY

Specifications

Cuff Material Pigskin Leather
Color Yellow
Hand Specific
Material Grain Cowhide Leather
Size Small
For Use With (Application) Linesmen's Work
Glove Cuff Color Brown
Glove Cuff Style Gauntlet
Series LW2SPE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.