Learn More
Honeywell Salisbury™ Pro-Wear™ 8 cal/cm2 Arc Flash Protection Clothing Kits with Coveralls
Catalog No. 19139777 Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-139-777 Medium
19-139-778 Large
19-139-776 Small
19-139-779 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-139-777 Supplier Honeywell Salisbury™ Supplier No. SKCA8M
This item is not returnable. View return policy
Specifications
Arc Flash Rating 8 cal/cm²
Description Coveralls, AS1000HAT, AFHOOD, salisbury storage bags and safety Glasses
Certifications/Compliance NFPA 70E™ 2009, CE 0120
Size Medium