Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Salisbury™ PRO-WEAR™ Flash Protection Bib Overalls

Catalog No. 19138125F Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Arc Flash Rating:
100 cal/cm2
11 cal/cm2
20 cal/cm2
31 cal/cm2
40 cal/cm2
75 cal/cm2
Color:
Gray
Khaki
Navy
Royal Blue
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
36 product options available for selection
Product selection table with 36 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Arc Flash Rating Color Size
19-138-125F 31 cal/cm2 Royal Blue 3X-Large
19-138-131E 100 cal/cm2 Khaki 2X-Large
19-138-131A 100 cal/cm2 Khaki Small
19-138-131F 100 cal/cm2 Khaki 3X-Large
19-138-131C 100 cal/cm2 Khaki Large
19-138-131B 100 cal/cm2 Khaki Medium
19-138-131D 100 cal/cm2 Khaki X-Large
19-138-120C 11 cal/cm2 Navy Large
19-138-120F 11 cal/cm2 Navy 3X-Large
19-138-120A 11 cal/cm2 Navy Small
19-138-120B 11 cal/cm2 Navy Medium
19-138-120D 11 cal/cm2 Navy X-Large
19-138-120E 11 cal/cm2 Navy 2X-Large
19-138-123E 20 cal/cm2 Navy 2X-Large
19-138-123F 20 cal/cm2 Navy 3X-Large
19-138-123A 20 cal/cm2 Navy Small
19-138-123B 20 cal/cm2 Navy Medium
19-138-125E 31 cal/cm2 Royal Blue 2X-Large
19-138-123C 20 cal/cm2 Navy Large
19-138-123D 20 cal/cm2 Navy X-Large
19-138-125C 31 cal/cm2 Royal Blue Large
19-138-125B 31 cal/cm2 Royal Blue Medium
19-138-125D 31 cal/cm2 Royal Blue X-Large
19-138-125A 31 cal/cm2 Royal Blue Small
19-138-127B 40 cal/cm2 Gray Medium
19-138-127C 40 cal/cm2 Gray Large
19-138-127E 40 cal/cm2 Gray 2X-Large
19-138-127F 40 cal/cm2 Gray 3X-Large
19-138-127A 40 cal/cm2 Gray Small
19-138-127D 40 cal/cm2 Gray X-Large
19-138-129A 75 cal/cm2 Gray Small
19-138-129C 75 cal/cm2 Gray Large
19-138-129D 75 cal/cm2 Gray X-Large
19-138-129F 75 cal/cm2 Gray 3X-Large
19-138-129B 75 cal/cm2 Gray Medium
19-138-129E 75 cal/cm2 Gray 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 36 options available.
36 options
This item is not returnable. View return policy
NFPA 70E does not have a Hazard Risk Category above 40 cal/cm2. Working on levels above 40 cal/cm2 should be avoied because of blast hazards caused by arc flash.
Catalog No. 19-138-125F Supplier Honeywell Salisbury™ Supplier No. ACB3130RB3X
This item is not returnable. View return policy
NFPA 70E does not have a Hazard Risk Category above 40 cal/cm2. Working on levels above 40 cal/cm2 should be avoied because of blast hazards caused by arc flash.

11 cal/cm2 to 100 cal/cm2 ATPV ratings

  • 11 cal/cm2 to 75 cal/cm2 overalls made of arc flash resistant Indura™ Ultra Soft™ fabric
  • 100 cal/cm2 overalls made of TuffWeld™ and Q/9™ material layers
  • All overalls sewn with Nomex™ for added flame protection
  • Relaxed cut for greater mobility with bib front for additional protection
  • 30 in. inseam and adjustable gussets on pant cuffs
  • Integrated heavy-duty suspenders and adjustable hook and pile waist straps

Compliance

Meet ASTM F1506 and NFPA 70E standards.

TRUSTED_SUSTAINABILITY

Specifications

Closure Type Adjustable Hook and Pile Waist Straps
Color Royal Blue
Material Indura™ Ultra soft™ Fabric
Size 3X-Large
Arc Flash Rating 31 cal/cm2
For Use With (Application) Flash protection
Length (English) 59 in.
Length (English) Inseam 30 in.
Length (Metric) 150 cm
Size (English) Waist 64 in.
Size (Metric) Waist 163 cm
Certifications/Compliance ASTM F1506, NFPA 70E™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.