Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Personal Protective Equipment
- Safety Clothing
- Protective Overalls
- Heat and Fire Resistant Overalls
- Honeywell Salisbury™ PRO-WEAR™ Flash Protection Bib Overalls
Honeywell Salisbury™ PRO-WEAR™ Flash Protection Bib Overalls
Catalog No. 19138127C Shop All WH Salisbury & Co. Products
Change view
Click to view available options
Arc Flash Rating:
100 cal/cm2
11 cal/cm2
20 cal/cm2
31 cal/cm2
40 cal/cm2
75 cal/cm2
Color:
Gray
Khaki
Navy
Royal Blue
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
36 product options available for selection
| Catalog No. | Arc Flash Rating | Color | Size |
|---|---|---|---|
| 19-138-127C | 40 cal/cm2 | Gray | Large |
| 19-138-131E | 100 cal/cm2 | Khaki | 2X-Large |
| 19-138-131A | 100 cal/cm2 | Khaki | Small |
| 19-138-131F | 100 cal/cm2 | Khaki | 3X-Large |
| 19-138-131C | 100 cal/cm2 | Khaki | Large |
| 19-138-131B | 100 cal/cm2 | Khaki | Medium |
| 19-138-131D | 100 cal/cm2 | Khaki | X-Large |
| 19-138-120C | 11 cal/cm2 | Navy | Large |
| 19-138-120F | 11 cal/cm2 | Navy | 3X-Large |
| 19-138-120A | 11 cal/cm2 | Navy | Small |
| 19-138-120B | 11 cal/cm2 | Navy | Medium |
| 19-138-120D | 11 cal/cm2 | Navy | X-Large |
| 19-138-120E | 11 cal/cm2 | Navy | 2X-Large |
| 19-138-123E | 20 cal/cm2 | Navy | 2X-Large |
| 19-138-123F | 20 cal/cm2 | Navy | 3X-Large |
| 19-138-123A | 20 cal/cm2 | Navy | Small |
| 19-138-123B | 20 cal/cm2 | Navy | Medium |
| 19-138-125F | 31 cal/cm2 | Royal Blue | 3X-Large |
| 19-138-125E | 31 cal/cm2 | Royal Blue | 2X-Large |
| 19-138-123C | 20 cal/cm2 | Navy | Large |
| 19-138-123D | 20 cal/cm2 | Navy | X-Large |
| 19-138-125C | 31 cal/cm2 | Royal Blue | Large |
| 19-138-125B | 31 cal/cm2 | Royal Blue | Medium |
| 19-138-125D | 31 cal/cm2 | Royal Blue | X-Large |
| 19-138-125A | 31 cal/cm2 | Royal Blue | Small |
| 19-138-127B | 40 cal/cm2 | Gray | Medium |
| 19-138-127E | 40 cal/cm2 | Gray | 2X-Large |
| 19-138-127F | 40 cal/cm2 | Gray | 3X-Large |
| 19-138-127A | 40 cal/cm2 | Gray | Small |
| 19-138-127D | 40 cal/cm2 | Gray | X-Large |
| 19-138-129A | 75 cal/cm2 | Gray | Small |
| 19-138-129C | 75 cal/cm2 | Gray | Large |
| 19-138-129D | 75 cal/cm2 | Gray | X-Large |
| 19-138-129F | 75 cal/cm2 | Gray | 3X-Large |
| 19-138-129B | 75 cal/cm2 | Gray | Medium |
| 19-138-129E | 75 cal/cm2 | Gray | 2X-Large |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 36 options available.
This item is not returnable.
View return policy
NFPA 70E does not have a Hazard Risk Category above 40 cal/cm2. Working on levels above 40 cal/cm2 should be avoied because of blast hazards caused by arc flash.
Catalog No.
19-138-127C
Supplier
Honeywell Salisbury™
Supplier No.
ACB4030GYL
This item is not returnable.
View return policy
NFPA 70E does not have a Hazard Risk Category above 40 cal/cm2. Working on levels above 40 cal/cm2 should be avoied because of blast hazards caused by arc flash.
Description
- 11 cal/cm2 to 75 cal/cm2 overalls made of arc flash resistant Indura™ Ultra Soft™ fabric
- 100 cal/cm2 overalls made of TuffWeld™ and Q/9™ material layers
- All overalls sewn with Nomex™ for added flame protection
- Relaxed cut for greater mobility with bib front for additional protection
- 30 in. inseam and adjustable gussets on pant cuffs
- Integrated heavy-duty suspenders and adjustable hook and pile waist straps
Compliance
Meet ASTM F1506 and NFPA 70E standards.
Specifications
Specifications
| Closure Type | Adjustable Hook and Pile Waist Straps |
| Color | Gray |
| Material | Indura™ Ultra soft™ Fabric |
| Size | Large |
| Arc Flash Rating | 40 cal/cm2 |
| For Use With (Application) | Flash protection |
| Length (English) | 56 in. |
| Length (English) Inseam | 30 in. |
| Length (Metric) | 142 cm |
| Length (Metric) Inseam | 76.2 cm |
| Size (English) Waist | 52 in. |
| Size (Metric) Waist | 132 cm |
| Certifications/Compliance | ASTM F1506, NFPA 70E™ |
| Show More Show Less |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?