Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Salisbury™ PRO-WEAR™ Flash Protection Coats

Catalog No. 19138122C Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Color:
Gray
Khaki
Navy
Royal Blue
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Arc Flash Rating:
100 cal/cm²
12 cal/cm²
20 cal/cm²
31 cal/cm²
40 cal/cm²
75 cal/cm²
36 product options available for selection
Product selection table with 36 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size Arc Flash Rating
19-138-122C Navy Large 20 cal/cm²
19-138-130E Khaki 2X-Large 100 cal/cm²
19-138-130F Khaki 3X-Large 100 cal/cm²
19-138-130B Khaki Medium 100 cal/cm²
19-138-130C Khaki Large 100 cal/cm²
19-138-119F Navy 3X-Large 12 cal/cm²
19-138-119E Navy 2X-Large 12 cal/cm²
19-138-130A Khaki Small 100 cal/cm²
19-138-119C Navy Large 12 cal/cm²
19-138-119B Navy Medium 12 cal/cm²
19-138-130D Khaki X-Large 100 cal/cm²
19-138-119D Navy X-Large 12 cal/cm²
19-138-119A Navy Small 12 cal/cm²
19-138-122F Navy 3X-Large 20 cal/cm²
19-138-122D Navy X-Large 20 cal/cm²
19-138-122E Navy 2X-Large 20 cal/cm²
19-138-122B Navy Medium 20 cal/cm²
19-138-122A Navy Small 20 cal/cm²
19-138-124E Royal Blue 2X-Large 31 cal/cm²
19-138-124B Royal Blue Medium 31 cal/cm²
19-138-124F Royal Blue 3X-Large 31 cal/cm²
19-138-124D Royal Blue X-Large 31 cal/cm²
19-138-126E Gray 2X-Large 40 cal/cm²
19-138-124C Royal Blue Large 31 cal/cm²
19-138-124A Royal Blue Small 31 cal/cm²
19-138-126F Gray 3X-Large 40 cal/cm²
19-138-126C Gray Large 40 cal/cm²
19-138-126B Gray Medium 40 cal/cm²
19-138-128C Gray Large 75 cal/cm²
19-138-126A Gray Small 40 cal/cm²
19-138-128E Gray 2X-Large 75 cal/cm²
19-138-126D Gray X-Large 40 cal/cm²
19-138-128F Gray 3X-Large 75 cal/cm²
19-138-128B Gray Medium 75 cal/cm²
19-138-128A Gray Small 75 cal/cm²
19-138-128D Gray X-Large 75 cal/cm²
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 36 options available.
36 options
This item is not returnable. View return policy
NFPA 70E does not have a Hazard Risk Category above 40 cal/cm2. Working on levels above 40 cal/cm2 should be avoied because of blast hazards caused by arc flash.
Catalog No. 19-138-122C Supplier Honeywell Salisbury™ Supplier No. ACC2032BLL
This item is not returnable. View return policy
NFPA 70E does not have a Hazard Risk Category above 40 cal/cm2. Working on levels above 40 cal/cm2 should be avoied because of blast hazards caused by arc flash.

11 cal/cm2 to 100 cal/cm2 ATPV ratings

  • 8 cal/cm2 to 75 cal/cm2 coats made from arc-flash resistant Indura™ Ultra Soft™ fabric
  • 100 cal/cm2 coats made from TuffWeld™ and Q/9™ material layers
  • All coats sewn with Nomex™ thread for added flame protection
  • 32 in. long coats feature expansion back for added comfort, FR hook and pile storm flap, and Nomex wristlets
  • Dual-stage front closure with high-temperature plastic zipper on 31 to 100cal/cm2 coats

Intended for use with Salisbury PRO-HOODS™.

Compliance

Meet current ASTM F1506 and NFPA 70E standards.

TRUSTED_SUSTAINABILITY

Specifications

Size Large
Material Indura™ Ultra soft™ Fabric
Color Navy
Closure Type FR Hook and Loop
Arc Flash Rating 20 cal/cm²
Certifications/Compliance ASTM F1506, NFPA 70E™, CE 0120
Size (English) Chest 42 to 44 in.
Length (English) 32 in.
Length (Metric) 81.28 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.