Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Personal Protective Equipment
- Safety Clothing
- Coats, Jackets, and Outerwear
- Heat and Fire Resistant Jackets
- Honeywell Salisbury™ PRO-WEAR™ Flash Protection Coats
Honeywell Salisbury™ PRO-WEAR™ Flash Protection Coats
Catalog No. 19138130A Shop All WH Salisbury & Co. Products
Change view
Click to view available options
Color:
Gray
Khaki
Navy
Royal Blue
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Arc Flash Rating:
100 cal/cm²
12 cal/cm²
20 cal/cm²
31 cal/cm²
40 cal/cm²
75 cal/cm²
36 product options available for selection
| Catalog No. | Color | Size | Arc Flash Rating |
|---|---|---|---|
| 19-138-130A | Khaki | Small | 100 cal/cm² |
| 19-138-130E | Khaki | 2X-Large | 100 cal/cm² |
| 19-138-130F | Khaki | 3X-Large | 100 cal/cm² |
| 19-138-130B | Khaki | Medium | 100 cal/cm² |
| 19-138-130C | Khaki | Large | 100 cal/cm² |
| 19-138-119F | Navy | 3X-Large | 12 cal/cm² |
| 19-138-119E | Navy | 2X-Large | 12 cal/cm² |
| 19-138-119C | Navy | Large | 12 cal/cm² |
| 19-138-119B | Navy | Medium | 12 cal/cm² |
| 19-138-130D | Khaki | X-Large | 100 cal/cm² |
| 19-138-119D | Navy | X-Large | 12 cal/cm² |
| 19-138-119A | Navy | Small | 12 cal/cm² |
| 19-138-122F | Navy | 3X-Large | 20 cal/cm² |
| 19-138-122D | Navy | X-Large | 20 cal/cm² |
| 19-138-122C | Navy | Large | 20 cal/cm² |
| 19-138-122E | Navy | 2X-Large | 20 cal/cm² |
| 19-138-122B | Navy | Medium | 20 cal/cm² |
| 19-138-122A | Navy | Small | 20 cal/cm² |
| 19-138-124E | Royal Blue | 2X-Large | 31 cal/cm² |
| 19-138-124B | Royal Blue | Medium | 31 cal/cm² |
| 19-138-124F | Royal Blue | 3X-Large | 31 cal/cm² |
| 19-138-124D | Royal Blue | X-Large | 31 cal/cm² |
| 19-138-126E | Gray | 2X-Large | 40 cal/cm² |
| 19-138-124C | Royal Blue | Large | 31 cal/cm² |
| 19-138-124A | Royal Blue | Small | 31 cal/cm² |
| 19-138-126F | Gray | 3X-Large | 40 cal/cm² |
| 19-138-126C | Gray | Large | 40 cal/cm² |
| 19-138-126B | Gray | Medium | 40 cal/cm² |
| 19-138-128C | Gray | Large | 75 cal/cm² |
| 19-138-126A | Gray | Small | 40 cal/cm² |
| 19-138-128E | Gray | 2X-Large | 75 cal/cm² |
| 19-138-126D | Gray | X-Large | 40 cal/cm² |
| 19-138-128F | Gray | 3X-Large | 75 cal/cm² |
| 19-138-128B | Gray | Medium | 75 cal/cm² |
| 19-138-128A | Gray | Small | 75 cal/cm² |
| 19-138-128D | Gray | X-Large | 75 cal/cm² |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 36 options available.
This item is not returnable.
View return policy
NFPA 70E does not have a Hazard Risk Category above 40 cal/cm2. Working on levels above 40 cal/cm2 should be avoied because of blast hazards caused by arc flash.
Catalog No.
19-138-130A
Supplier
Honeywell Salisbury™
Supplier No.
ACC10032TWS
This item is not returnable.
View return policy
NFPA 70E does not have a Hazard Risk Category above 40 cal/cm2. Working on levels above 40 cal/cm2 should be avoied because of blast hazards caused by arc flash.
Description
- 8 cal/cm2 to 75 cal/cm2 coats made from arc-flash resistant Indura™ Ultra Soft™ fabric
- 100 cal/cm2 coats made from TuffWeld™ and Q/9™ material layers
- All coats sewn with Nomex™ thread for added flame protection
- 32 in. long coats feature expansion back for added comfort, FR hook and pile storm flap, and Nomex wristlets
- Dual-stage front closure with high-temperature plastic zipper on 31 to 100cal/cm2 coats
Intended for use with Salisbury PRO-HOODS™.
Compliance
Meet current ASTM F1506 and NFPA 70E standards.
Specifications
Specifications
| Size | Small |
| Material | TuffWeld, Q/9 |
| Color | Khaki |
| Closure Type | Zipper |
| Arc Flash Rating | 100 cal/cm² |
| Certifications/Compliance | ASTM F1506, NFPA 70E™, CE 0120 |
| Size (English) Chest | 34 to 36 in. |
| Size (Metric) Chest | 86.36 to 91.44 cm |
| Length (English) | 32 in. |
| Length (Metric) | 81.28 cm |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?