Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Salisbury™ Roll Blankets
Protection from low-voltage electrical hazards
Supplier: Honeywell Salisbury™ RLBPVC1
Description
- Made from a high-strength fabric reinforced Type II rubber
- All classes of blankets come in 3 × 30 ft. rolls that can be easily custom cut to fit each application
- Self extinguishing and highly resistant to punctures, tears, oil and ozone
- Unique colors make each class easy to identify and highly visible in the workplace
- Class I clear PVC blanket allows complete visibility and provides the necessary insulating properties to meet ASTM F1742 standards
- Blanket clamp pins, made of fiberglass reinforced nylon with molded rubber tips, can be used to hold blankets in place
Specifications
Clear | |
ASTM F2320 | |
360 x 36 in. (914 x 91.44 cm) | |
360 in. | |
7500 V | |
16 kg | |
91.44 cm |
PVC | |
Class 1 | |
Protection from low voltage electrical hazards, easily cut to fit | |
914 cm | |
36 lb. | |
36 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction