Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Salisbury™ RUB-OUT™ Towelettes

Catalog No. 19138427 Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Packaging:
100 Single Towelettes in Carton
Tub of 60 towelettes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
19-138-427 100 Single Towelettes in Carton
19-138-426 Tub of 60 towelettes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-138-427 Supplier Honeywell Salisbury™ Supplier No. 1461

Safe to use with insulating rubber gloves

  • Non-petroleum-based hand cleaner dissolves and removes grease, oil, ink, tar, paint and more
  • Safe to use with natural rubber or ozone-resistant SALCOR™ rubber
  • Cleans with or without water
  • Contains natural skin conditioners and leaves a fresh citrus scent

TRUSTED_SUSTAINABILITY

Specifications

Product Type Rub-Out™ Towelettes
For Use With (Application) Dissolves grease, oil, ink, tar, paint, cleans with or without water, dissolves grease, oil, ink, tar, paint
For Use With (Equipment) Rubber gloves
Packaging 100 Single Towelettes in Carton
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.