Learn More
Invitrogen™ Sall1 Monoclonal Antibody (NRNSTNX), eBioscience™

Description
Description: This NRNSTNX monoclonal antibody recognizes mouse Sall1. This clone has been reported to be successfully used in flow cytometry, native gel electrophoresis followed by western blot and immunohistochemistry of frozen tissue. Applications Reported: This NRNSTNX antibody has been reported for use in native gel electrophoresis followed by western blot and in immunohistochemical staining. Applications Tested: This NRNSTNX antibody has been tested by western blot (native) of mouse brain. This may be used at less than or equal to 5 μg/mL. This clone is not recommended for detection of denatured Sall1. Applications Tested: This NRNSTNX antibody has been tested by immunohistochemical staining of acetone fixed frozen sections from mouse brain. This may be used at less than or equal to 1.25 μg/mL. (Fluorochrome-conjugated NRNSTNX is recommended for use in flow cytometry).
Specifications
Specifications
| Antigen | Sall1 |
| Applications | Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | NRNSTNX |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | SALL1 |
| Gene Accession No. | Q9ER74 |
| Gene Alias | epididymis secretory protein Li 89; HEL-S-89; HSal1; Msal-3; SAL1; Sal-1; sal-3; SALL1; sal-like 1; sal-like 1 (Drosophila); sal-like protein 1; spalt like transcription factor 1; spalt-like transcription factor 1; TBS; Zinc finger protein 794; zinc finger protein SALL1; Zinc finger protein Spalt-1; zinc finger protein Spalt-3; ZNF794 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.