Learn More
Biocare Medical, LLC SALL4 Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CM384A
Encompass_Preferred
SALL4 is required for the maintenance of embryonic stem cell pluripotency by modulating Oct4. Studies support SALL4 as a sensitive and specific marker for seminomas and ovarian primitive germ-cell tumors. Studies have demonstrated that over 90% of tumor cells in intratubular germ-cell neoplasias and embryonal carcinomas show strong SALL4 staining. In addition, 100% of 31 yolk sac tumors (5 pediatric and 26 postpubertal) showed strong positive SALL4 staining of tumor cells, but were negative for Oct4. SALL4 antibody is a promising pan germ-cell marker, with studies showing that it is superior to PLAP and Oct4 antibodies.
Catalog No. 50-826-79
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.