Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Salmeterol Xinafoate, Beta2 Adrenoceptor Agonist, Solid, 10 mg

Catalog No. 502707286
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7286 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7286 Supplier Abcam Supplier No. AB12077110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective, long acting β2 adrenoceptor agonist that inhibits histamine and leukotriene release.

  • Purity: >96%
  • Molecular Weight: 603.7 Da
  • CAS Number: 94749-08-3
  • Properties: Selective, long acting β2 adrenoceptor agonist
  • Activity: Inhibits release of histamine, leukotrienes, prostaglandin D2 and GM-CSF in concentration-dependent manner; active in vitro and in vivo
  • Storage: Store at Ambient
  • Form: Solid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Beta2 Adrenoceptor
Content And Storage Store at ambient temperature.
Form Solid
Shipping Condition Ambient
Quantity 10 mg
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.