Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Salmeterol Xinafoate, Beta2 Adrenoceptor Agonist, Solid, 50 mg

Catalog No. 502707285
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7285 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7285 Supplier Abcam Supplier No. AB12077150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective, long acting β2 adrenoceptor agonist that inhibits the release of histamine and leukotrienes.

  • Molecular weight: 603.7 Da
  • Purity: >96%
  • Selective, long acting β2 adrenoceptor agonist
  • Inhibits release of histamine, leukotrienes, prostaglandin D2 and GM-CSF in a concentration-dependent manner
  • Active in vitro and in vivo

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Beta2 Adrenoceptor
Shipping Condition Ambient
Content And Storage Store at ambient temperature.
Form Solid
Quantity 50 mg
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.