Learn More
Kingfisher Biotech Inc Atlantic Salmon TNF alpha Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1327AS025
Encompass_Preferred
The Atlantic Salmon TNF alpha yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Atlantic Salmon TNF alpha applications are for cell culture, ELISA standard, and Western Blot Control. Atlantic Salmon TNF alpha yeast-derived recombinant protein can be purchased in multiple sizes. Atlantic Salmon TNF alpha Specifications: (Molecular Weight: 16.9 kDa) (Amino Acid Sequence: AAIHLEGEYN PNLTADTVQW RKDDGQAFSQ GGFELQGNQI LIPHTGLFFV YSQASFRVKC NGPGENTTPL SHVIWRYSDS IGVNANLLSG VRSVCQRNYG DAESNIGEAW YNAVYLGAVF QLNEGDKLWT ETNRLTNVEP EQGKNFFGVF EL (152)) (Gene ID: 100136457). For research use only. Made in the USA
Catalog No. 50-253-3379
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.