Learn More
ATCC Salmonella enterica; serovar Typhimurium LT2; genomic DNA
Supplier:  ATCC 700720D5
Encompass
Genomic DNA isolated from Salmonella enterica subsp. enterica serovar Typhimurium strain LT2. This whole-genome sequenced product can be used in assay development and zoonotic disease research.
Catalog No. 50-238-3703