Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Salmonella enterica subsp. enterica serovar Minnesota Sal60
SDP

Catalog No. 12111415 Shop All Microbiologics Products
Change view
Click to view available options
Format:
UV-BioTag Swab
UV-BioTag Vial
Description:
Derived from FDA SAL5698
Derived from FDA SAL5699
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Description
12-111-415 UV-BioTag Vial Derived from FDA SAL5698
12-111-416 UV-BioTag Swab Derived from FDA SAL5699
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-111-415 Supplier Microbiologics™ Supplier No. 01228UVV
Add to Cart
Add to Cart
This item is not returnable. View return policy

UV-BioTAG microorganisms contain a Green Fluorescent Protein (GFP) marker that produces fluorescence of the culture visible under UV light. This allows for easy differentiation of these control strains from contaminants. Kits include six vials or swabs, each containing one lyophilized pellet of an individual microorganism strain.

It’s not uncommon for food samples to inadvertently become contaminated with control strains used in the laboratory. This leads to false-positive results which can have severe financial and regulatory implications. UV-BioTAG, our line of reference strains containing green fluorescent protein (GFP) markers, make it easy to distinguish standard microorganism strains from laboratory control strains. Optimized for highly visible fluorescence, UV-BioTAG strains provide better stability because the GFP marker is integrated into the chromosome rather than the plasmid. With UV-BioTAG, you can quickly and reliably establish whether a positive test result can be traced back to a control strain cross-contamination issue, or if it is from another source.

Specifications

Biosafety Level 2
Content And Storage 2°C to 8°C
Description Derived from FDA SAL5698
Format UV-BioTag Vial
Manufacturing Origin United States
Product Type Quality Control
For Use With (Application) Food and Food Ingredients: Pathogen Testing
Packaging Type Plastic Tube
Quantity 6/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.