Learn More
Description
PA1-20811 detects Salmonella, and is polyvalent for all 'O & H' Salmonella antigens.
Specifications
Specifications
| Antigen | Salmonella |
| Applications | Immunoprecipitation, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | Salmonellae |
| Host Species | Rabbit |
| Immunogen | Mixture of S. enteriditis, S. typhimurium & S. heidelburg. |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
