Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Samarium(III) chloride, ultra dry, 99.9% (REO)

Catalog No. AA3580409
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3580409 10 g
AA3580404 2 g
AA3580418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3580409 Supplier Thermo Scientific Chemicals Supplier No. 03580409
Add to Cart

CAS: 10361-82-7 | Cl3Sm | 256.71 g/mol

This compound named Samarium(III) chloride has few practical applications but is used in laboratories for research on new compounds of samarium. Samarium(III) chloride is used for the preparation of samarium metal, which has a variety of uses, notably in magnets. Anhydrous SmCl3 is mixed with sodium chloride or calcium chloride to give a low melting point eutectic mixture. Electrolysis of this molten salt solution gives the free metal. Samarium(III) chloride can also be used as a starting point for the preparation of other samarium salts. The anhydrous chloride is used to prepare organometallic compounds of samarium, such as bis(pentamethylcyclopentadienyl)alkylsamarium(III) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-82-7
Molecular Formula Cl3Sm
Molecular Weight (g/mol) 256.71
MDL Number MFCD00011235
InChI Key BHXBZLPMVFUQBQ-UHFFFAOYSA-K
IUPAC Name samarium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Sm+3]

Specifications

Boiling Point (decomposition)
Melting Point 678°C
Physical Form Powder
Quantity 10 g
Merck Index 14,8347
Solubility Information It is soluble in water.
Formula Weight 256.71
Percent Purity 99.9%
Packaging Ampule
Sensitivity Hygroscopic
Density 4.46 g/mL at 16°C
Assay (REO)
Chemical Name or Material Samarium(III) chloride, Ultra dry
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 233-797-0
RTECSNumber VP2625000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.