Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sambucus Nigra (Elderberry Bark) Lectin (SNA, EBL), fluorescein (FITC)

Catalog No. L32479
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L32479 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L32479 Supplier Invitrogen™ Supplier No. L32479
  • $258.00
    $211.56 / Each of 1

    Save $46.44

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Bright-green SNA-fluorophor conjugate

  • Sambucus Nigra (Elderberry Bark) Lectin (SNA, EBL), fluorescein (FITC), is a bright-green SNA-fluorophor conjugate. SNA belongs to the group of type 2 ribosome-inactivating proteins and is composed of an A-chain with enzymatic activity and a B-chain with carbohydrate-binding activity. SNA can induce apoptosis through the caspase-dependent pathway at low concentrations.
  • This effect depends on the carbohydrate binding of the B-chain and is independent from the catalytic activity of the A-chain. In colon cancers, it has been reported that 90% of carcinomas express increased levels of sialic acid, linked to galactose residues on the surface of tumor cells, that is detectable with SNA.
  • Very bright FITC conjugate detected with traditional FITC/GFP filter set, Ex/Em: 495/515 nm
  • Applications: immunofluorescence, glycobiology
  • Sugar specificity: sialic acid
  • Inhibiting/eluting sugar: 500 mM lactose
  • Blood group specificity: non-specific, A > B, O
  • Unconjugated lectin molecular weight: 140 KDa
  • Immunofluorescence working concentration: 5–20 μg/mL
  • SNA-FITC is provided at a 2 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide. 0.1 mM Ca2+
  • A precipitate may form during storage. This does not have a significant adverse effect on the product. If a precipitate forms upon long-term storage at 4°C, warm to 37°C and centrifuge before use.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 mg
Source Elderberry
Conjugate Fluorescein
Content And Storage When stored at or below 2°C to 8°C in the dark, the product is stable for 6 months after reciept.
Label Type Classic Dyes
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Solution
Protein Subtype Lectins

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.