Learn More
Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers
Description
Prevent bio-hazardous spills with the proprietary, all plastic, leak-resistant seal of Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 120 mL (4 oz.) 53mm Specimen Containers. Wide-mouth containers are ideally suited for urine collection from women during pregnancy and for older or overweight patients. These stackable, shatter-resistant containers are manufactured from virgin polypropylene and feature the Bio-Tite™ system. They are tested at varying pressures, making them ideal for use in various transport systems.
Features:
- 120mL (4 oz.) 53mm
- Bio-Tite caps create leak-resistant seal for superior safety and handling
- Dual-threaded caps create improved sealing
- Sterilized by gamma irradiation, which eliminates potential contamination
- Top and bottom positive grip knurls are easy to grip and open using latex gloves
- Stackable containers are easy to store, saving lab space
- Virgin polypropylene containers allow higher clarity to see samples, and will not become brittle
- Sterile indicator tab assures tamper evidence
- Bilingual English and Spanish label facilitates accurate usage
Ordering Information:
Case of 300
Specifications
Specifications
| Product Type | Specimen Container |
| Closure Included | Yes |
| Material | Virgin Polypropylene |
| Shape | Cylindrical |
| Sterility | Sterile |
| Capacity (Metric) | 120 mL |
| Closure Color | White |
| Closure Size | 53 mm |
| Closure Type | Bio-Tite Lid |
| Description | Cap and container pre-assembled, includes sodium thiosulfate tablet |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.