Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAMM50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100325
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100325 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100325 Supplier Invitrogen™ Supplier No. PA5100325
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SAMM50.

SAMM50 is a component of the sorting and assembly machinery (SAM) complex of the outer mitochondrial membrane. The SAM complex has a role in integrating beta-barrel proteins into the outer mitochondrial membrane (Humphries et al, 2005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAMM50
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Samm50
Gene Accession No. Q6AXV4, Q8BGH2, Q9Y512
Gene Alias 1110030L07Rik; CGI-51; OMP85; SAM50; SAMM50; SAMM50 sorting and assembly machinery component; sorting and assembly machinery 50kDa; sorting and assembly machinery component 50 homolog; sorting and assembly machinery component 50 homolog (S. cerevisiae); sorting and assembly machinery component 50 homolog A; TOB55; Transformation-related gene 3 protein; TRG3; TRG-3; YNL026W
Gene Symbols Samm50
Host Species Rabbit
Immunogen A synthesized peptide derived from human SAMM50(Accession Q9Y512), corresponding to amino acid residues A334-Q384.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25813, 300111, 68653
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.