Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Sample Activation Kit 1
SDP

Catalog No. DY010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY010 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY010 Supplier R&D Systems™ Supplier No. DY010

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sample Activation Kit 1 from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Sample Activation Kit 1 quantifies multi-species Buffers, Chelators and Reagents.

Specifications

Product Line R&D Systems
For Use With (Application) ELISA
Quantity 1 Kit
Storage Requirements Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.