Learn More
Market Lab Magazine Rack for Roche, Hitachi, and Centaur, 11.25""""L x 5""""W x 2""""H, Slot Dimensions: 4.75""""L x 0.875""""W x 1""""H
Supplier:  Market Lab 1015
Encompass_Preferred
Are you frustrated after another magazine rack just tipped over? Check out this unique solution from MarketLab. These racks are designed specifically to keep Centaur, Roche, and Hitachi magazines upright, making your job easier and faster.
Catalog No. NC1479192
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.