Learn More
Description
- Supplied with lid
- Flat bottom
Specifications
Specifications
| Product Type | Analyzer Cup |
| Volume (Metric) | 25 mL |
| Color | Clear |
| Material | Polystyrene |
| For Use With (Equipment) | Coulter Counter analyzer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
