Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Sample Cups for Roche Cobas Mira

Catalog No. 21200124 Shop All Globe Scientific Products
Change view
Click to view available options
Color:
Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-200-124 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-200-124 Supplier Globe Scientific Supplier No. 6500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Designed for use on automated instrumentation including the Cobas Mira, Bio, Fara and Horiba ABX analyzers. Globe Scientific Sample Cups for Roche Cobas Mira are constructed of high density polyethylene (HDPE) and feature an attached snap cap.

  • For use on automated instrumentation including the Cobas Mira, Bio, Fara and Horiba ABX analyzers
  • Attached snap cap
  • High Density Polyethylene (HDPE)
  • Green (not pictured)

Specifications

Color Green
Material HDPE
Product Type Sample Cup
For Use With (Equipment) Cobas Mira, Bio, Fara and Horiba ABX Analyzers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.