Learn More
MilliporeSigma™ Supelco™ Sample Diffusion Discs for LC columns
A 20 μm polyethylene disc sealed to an acrylic ring. Installed on top of column packing in an open-ended column, it protects the packing from disruption.
$41.75
Specifications
For Use With (Equipment) LC Columns
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
11-101-1925
View Documents
MilliporeSigma™ Supelco™
S72711EA
Column I.D. 2.5 cm
Each of 1 for $41.75
Only null left
Add to Cart
Description
Description
Specifications
Specifications
LC Columns
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.