Learn More
R&D Systems™ Sample Diluent SD20 - 8 mL
Specifications
Specifications
| Quantity | 3 Vial |
| Storage Requirements | Store unopened at 2-8 °C. Stable for up to 30 days after opening when stored at 2-8 °C. |