Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sample Grinding Kit

Catalog No. 45001731 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50 Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-731 50 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-731 Supplier Cytiva Supplier No. 80648337
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Microcentrifuge tubes with grinding resin, 50; Pestles, 50

Disrupts small tissue and cell samples for protein extraction

  • Uses 1.5mL microcentrifuge tubes, grinding resin, and disposable pestles
  • Process up to 100mg of sample per tube in 10 minutes

TRUSTED_SUSTAINABILITY

Specifications

Product Type Sample grinding kit
Quantity 50 Samples
Includes 1.5mL Microcentrifuge tubes prepacked with grinding Resin, disposable pestles
Content And Storage Room Temperature
For Use With (Application) Mechanical breaking up of cells or tissue releasing the to tal Protein content
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.