Learn More
Thermo Scientific SAMPLE INTRO SPARES KIT
Supplier:  Thermo Scientific 842312051751
Encompass
Sample Introduction Spares Kit; Consumables kit containing a replacement sample introduction tubing &; connector kit, check valve assembly, sampling O-ring kit and a sampling; probe with capillary sleeving; Compatible with all iCAP 6000 Series Spectrometers.
Catalog No. NC9012576
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.