Learn More
Advanced Instruments Micro-Sample Test Kit
SDP
Catalog No. 22047530 Shop All Advanced Instruments Products
Change view
Click to view available options
For Use With (Equipment):
Advanced Instruments Osmo1 and 3320 Single-Sample Micro Osmometers
Catalog No. For Use With (Equipment)
22-047-530 Advanced Instruments Osmo1 and 3320 Single-Sample Micro Osmometers
1 options
Catalog No. 22-047-530 Supplier Advanced Instruments Supplier No. 133800
Add to Cart
Add to Cart
The Advanced Instruments Micro-Sample Test Kit ensures accurate and precise results with the Advanced Instruments Osmo1 and 3320 Single-Sample Micro-Osmometers.
  • 500 chamber cleaners
  • 500 sampler tips
  • 1 plunger wire
Specifications
Product Type Chamber Cleaners, Sampler Tips, Plunger Wire
For Use With (Equipment) Advanced Instruments Osmo1 and 3320 Single-Sample Micro Osmometers
Disposable Single-use
For Use With (Application) Osmolality testing of aqueous solutions