Learn More
Kitted Lab Technologies Sample Line - Bioreactor Kit 20/box
Supplier:  Kitted Lab Technologies K01003
Encompass
KLT's Sample line with swab valve. Plugged ends for sterile environment. Can easily be spliced to existing bioreactor set-ups. Incldes 1/4" pinch clamp on advantaflex tubing. Assembled and packaged in Class 9 clean room. Individually packaged with 20 lines per box. KLT specializes in Custom kit configurations and would be happy to discuss your specific lab set-up needs.
Catalog No. NC2086252
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.